Learn More
Waters Corp Waters Corporation Adapter, PEEK, Female 1/4-28 to Male 10-32
Supplier:  Waters Corp 700004597
Encompass_Preferred
This adapter is a pressure-relief device designed to protect the fluidics valve's rotor and stator. It functions by breaking the seal to the stainless steel tube if the pressure exceeds approximately 1500 psi.
  • Made from Tan PEEK material.
  • Features female 1/4-28 to male 10-32 connection types.
  • Protects the fluidics valve's rotor and stator.
  • Breaks seal to stainless steel tube if pressure exceeds approximately 1500 psi.
Catalog No. 50-793-023