Learn More
Electron Microscopy Sciences Evactron Adapter Flange JEOL, 100x355 mm, NO O-RING
SDP
Supplier:  Electron Microscopy Sciences 23002201
Encompass_Preferred
Evactron adaptor flanges are identified by physical dimensions of the blank plate in the SEM/FIB. ORID is a measurement of the inner diameter of the O ring.
Size: 100 X 355 mm
O-ring ID: NO O-RING
Mounting holes: 5 ASYM
Catalog No. 50-192-8255