Learn More
Beckman Coulter 16mm Diameter Tube Adapter Assembly for Multi-Disc, 2/pk
Supplier:  Beckman Coulter 359471
Encompass
Color: Purple. Tube Volumes: 12 mL. Max Tube Diameter: 16-mm. Number of Places in Adapter: 19. Number of Tubes in Rotor: 76.
Catalog No. NC1102647