Learn More
Chemglass Life Sciences #2 Neoprene Filter Adapter, Minum-Ware
SDP
Supplier:  Chemglass Life Sciences MW12102
Encompass_Preferred
#2 Neoprene Filter Adapter, Minum-Ware. Microscale neoprene filter adapters for use with MW-122 filtering flask. Adapters may be nested to permit use of funnels with a stem diameter as small as 6mm OD.
Catalog No. 50-871-931