Learn More
Chemglass Life Sciences PTFE adapter, GL80 to 24/40 outer joint - for GL80 wide-mouth recovery flasks
SDP
Supplier:  Chemglass Life Sciences CG1512X20
Encompass_Preferred
The Chemglass CG-1512-X-20 is a PTFE adapter that connects a GL80 threaded wide-mouth recovery flask to a 24/40 outer joint. It uses a PTFE envelope-type seal with an FDA-grade silicone gasket so that liquids and powders contact only glass or PTFE; it is suitable for vacuum applications and for handling viscous materials.
  • PTFE construction minimizes chemical contamination and interaction.
  • FDA-grade silicone gasket behind a PTFE envelope seal protects contents.
  • Connects GL80 glass thread to 24/40 outer joint for recovery flask configurations.
  • Designed for vacuum applications and handling viscous liquids or powders.
  • Easy to clean; limits exposure of contents to non-PTFE materials.
Catalog No. 50-273-3589