Learn More
Chemglass Life Sciences PFA Straight Adapter, 1/8" Compression Fitting to 1/4" NPT
SDP
Supplier:  Chemglass Life Sciences CG1975F16
Encompass_Preferred
PFA Straight Adapter, 1/8" Compression Fitting to 1/4" NPT. PFA straight adapter, compression to male NPT fittings are ideally suited for use in corrosive environments and chemical applications. The fitting design yields a leak-tight connection and requires no special tools to assemble. The connection is achieved by merely inserting the tubing into the fitting assembly and tightening the nut. Note: Fittings have a shoulder/step. Tubing can not pass through the entire fitting. See CG-1971-81 Series fittings for this type of application.
Catalog No. 50-143-616