Learn More
Beckman Coulter Adapter, 1/4-28 to M6
Supplier:  Beckman Coulter 342327
Encompass
The Beckman Coulter Microfuge 18 Centrifuge Rotor Adapter is designed to adapt 0.2mL and 0.5mL microcentrifuge tubes to fit in the Microfuge 18 Centrifuge. It allows the centrifuge to spin smaller volume tubes that normally would not fit. The rotor adapter is made of durable polypropylene and can hold up to 18 microcentrifuge tubes. It fits seamlessly into the Microfuge 18 rotor and is easy to load and unload tubes. This adapter expands the versatility of the Microfuge 18 for research requiring smaller volume tubes.
Catalog No. NC0268357