Learn More
Beckman Coulter Adapter, 1/4-28 to M6
Supplier:  Beckman Coulter 347539
Encompass_Preferred
The Beckman Coulter Microfuge 18 Centrifuge Rotor Adapter is designed to adapt 0.2ml, 0.5ml, and 1.5ml microcentrifuge tubes to the Microfuge 18 Centrifuge. It allows centrifugation of up to 18 tubes at a time. The rotor is made of durable polycarbonate and can reach speeds up to 13,300 rpm. This is an OEM part that fits the Microfuge 18 Centrifuge perfectly for high-speed microcentrifuge tube centrifugation.
Catalog No. NC1591790