Learn More
Starna Cells Inc ADAPTOR FOR CELL 23545Q5
SDP
Supplier:  Starna Cells Inc FCA5
Encompass_Preferred
Adaptor, Black Anodized, Exterior Size: 12.5W x 12.5 L x 45mmH, Interior Size: 7.5W x 7.5mm L, Fits Cell Type: 3-5, Aperature Size: 4.5W x 11mmH, Quantity Windows 3
Catalog No. NC9263381