Learn More
Electron Microscopy Sciences Evactron Adapter Flange JEOL, 75x98 mm
SDP
Supplier:  Electron Microscopy Sciences 23032201
Encompass_Preferred
Evactron adaptor flanges are identified by physical dimensions of the blank plate in the SEM/FIB. ORID is a measurement of the inner diameter of the O ring.
Size: 75x98 ARCHED, 1 SIDE
Mounting holes: 4 ASYM
Catalog No. 50-192-8258