Learn More
Santa Cruz Biotechnology Anti-ADAR1 Antibody (15.8.6), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC73408
Encompass
Anti-ADAR1 Antibody (15.8.6) is recommended for detection of native and recombinant ADAR1 of mouse, rat and human origin by WB, IP, IF and IHC(P); non cross-reactive with other members of ADAR family.
Catalog No. NC1600317