Learn More
Selleck Chemical LLC Adavivint S8761-25MG
SDP
Supplier:  Selleck Chemical LLC S876125MG
Encompass_Preferred
Adavivint (SM04690 Lorecivivint) is a potent and specific inhibitor of canonical Wnt signaling with an EC50 of 19 5 nM for inhibiting the TCF/LEF reporter It is 150- to 500-fold more potent than the other known Wnt inhibitors across multiple cellular assays
Catalog No. 50-000-24382