Learn More
Medchemexpress LLC Adavosertib | 955365-80-7 | 99.9% | 500.60 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10993R10MG
Encompass_Preferred
Adavosertib (Standard) is the analytical standard of Adavosertib, intended for research and analytical applications. It is a potent Wee1 inhibitor with an IC50 of 5.2 nM.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-003-44466