Learn More
Zymo Research Corporation ADB (Agarose Dissolving Buffer), 100 ml
SDP
Supplier:  Zymo Research Corporation D40011100
Encompass_Preferred
This buffer dissolves agarose gel slices for the recovery of DNA and facilitates efficient DNA binding and subsequent recovery.
Catalog No. 50-444-175