Learn More
Strem, An Ascensus Company CAS# 1160861-59-5. 500mg. 2-(Di-1-adamantylphosphino)-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl, min. 95% AdBrettPhos. MFCD23379935
SDP
Supplier:  Strem, An Ascensus Company 151138500MG
Encompass
CAS# 1160861-59-5. 500mg. 2-(Di-1-adamantylphosphino)-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl, min. 95% AdBrettPhos. MFCD23379935. Molecular Weight: 640.92. Molecular Formula: C43H61O2P. Color/form: off-white to pale yellow pwdr. Strem# 15-1138. http://www.strem.com/catalog/v/15-1138/
Catalog No. 50-201-6784