Learn More
Medchemexpress LLC HY-B1826 10mg Medchemexpress, Adefovir CAS:106941-25-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB182610MG
Encompass_Preferred
Medchemexpress, HY-B1826 10mg Adefovir CAS:106941-25-7 Adefovir (GS-0393) is an adenosine monophosphate analog antiviral agent that after intracellular conversion to Adefovir diphosphate inhibits HBV DNA polymerase. Adefovir has an IC50 of 0.7 μM against HBV in the HepG2.2.15 cell line. Adefovir has good antiviral activity against several viruses, including HBV and herpesviruses[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0166