Learn More
Cayman Chemical AdefovIr-d4 2.5mg
SDP
Supplier:  Cayman Chemical 313152.5 MG
Encompass_Preferred
An internal standard for the quantification of adefovir by GC- or LC-MS
Catalog No. 50-299-0939