Learn More
Selleck Chemical LLC Adefovir Dipivoxil 100mg 142340-99-6
SDP
Supplier:  Selleck Chemical LLC S1718100MG
Encompass_Preferred
Adefovir Dipivoxil is a reverse transcriptase inhibitor, used in the treatment of chronic hepatitis B virus (HBV). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-503-1