Learn More
Apexbio Technology LLC Ademetionine 29908-03-0 500mg
SDP
Supplier:  Apexbio Technology LLC B3513500
Encompass_Preferred
Ademetionine (CAS 29908-03-0) also termed S-adenosylmethionine (SAMe) is an endogenous metabolite involved as a methyl group donor in numerous cellular methylation processes including the methylation of DNA proteins catecholamines and phospholipids Mechanistically ademetionine modulates neurotransmission by elevating concentrations of brain monoamine neurotransmitters and enhancing muscarinic and -adrenergic receptor function Due to these neurochemical effects ademetionine has been explored in research contexts for psychiatric disorders such as depression and investigated for possible therapeutic roles in dementia AIDS-related myelopathy and ischemic brain injury
Catalog No. 50-000-63880