Learn More
Sigma Aldrich Fine Chemicals Biosciences Adenine 99100G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A8626100G
Encompass
Adenine is a purine nucleobase. It is part of DNA and RNA. Adenine is also a component of cofactors (NAD fAD) and signaling molecules (cAMP).
Catalog No. 50-290-3427