Learn More
Medchemexpress LLC Adenine hydrochloride | 2922-28-3 | MFCD00075782 | 98.0% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB0152A1G
Encompass_Preferred
Adenine hydrochloride is a purine nucleobase, crucial for DNA and RNA. It plays a significant role in cellular respiration, forming ATP and cofactors like NAD and FAD, and is involved in protein synthesis. This product is intended for research use only.
  • Component of DNA and RNA
  • Involved in cellular respiration and ATP formation
  • Plays a role in cofactor synthesis (NAD, FAD)
  • Supports protein synthesis
  • Can be used to induce chronic liver injury models
  • White to off-white solid appearance
Catalog No. 50-004-07235