Learn More
ATCC Adeno-associated virus 2; Strain: H
Supplier:  ATCC VR680
Encompass
Adeno-associated virus 2 strain H is propagated in 293 [HEK-293] cells (ATCC CRL-1573) and depends on the helper virus Human adenovirus 2 (ATCC VR-846) for replication. This strain was isolated from a culture of adenovirus type 12 strain 97838 in KB cells and has applications in gene therapy research.
Catalog No. 50-238-3744