Learn More
ATCC Adeno-associated virus 5; Strain: Original
Supplier:  ATCC VR1523
Encompass
Adeno-associated virus 5 is propagated in LLC-MK2 Derivative cells (ATCC CCL-7.1) with a helper adenovirus. This strain was isolated from a human clinical specimen.
Catalog No. 50-238-4102