Learn More
Medchemexpress LLC Adenosine-1'-13C | 201996-55-6 | 99.7% | 268.23 g·mol^-1 | C10H13N5O4 (1-13C) | 10 MG

Supplier: Medchemexpress LLC HYB0228S410MG
A stable isotope-labeled adenosine with the 13C label at the 1′ position, provided as a powder for analytical and tracer applications. It serves as an isotopic standard or tracer in biochemical, metabolic, and receptor studies where labeled nucleosides are required.
- Contains a single 13C label at the 1′ position for tracer and quantitative studies.
- High purity suitable for analytical use (reported purity ~99.66%).
- Soluble in DMSO (high concentration; ultrasonic assistance recommended).
- Supplied as a compact powder for long-term storage at recommended temperatures.
- Molecular weight approximately 268.23 g·mol^-1, compatible with mass spectrometry and NMR applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.