Learn More
Medchemexpress LLC Adenosine, 1-oxide | 146-92-9 | MFCD00037993 | 99.5% | 283.24 g/mol | C10H13N5O5 | 1 MG

Supplier: Medchemexpress LLC HY1020821MG
Adenosine N1-oxide is a nucleoside analog reported as an orally active anti-inflammatory agent that also promotes osteogenic and adipocyte differentiation. It is supplied as a white to off-white solid of high purity for use in research applications.
- Oral active anti-inflammatory agent.
- Promotes osteogenic and adipocyte differentiation in reported studies.
- High purity (99.5%) suitable for research experiments.
- Molecular formula C10H13N5O5; molecular weight 283.24 g/mol.
- Available in milligram-scale quantities for laboratory use.
- Store powder at -20 °C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.