Learn More
Medchemexpress LLC Adenosine 2'-monophosphate | 130-49-4 | MFCD00022827 | 99.9% | 347.22 g/mol | C10H14N5O7P | 5MG
SDP
Supplier:  Medchemexpress LLC HY1241515MG
Encompass_Preferred
Adenosine-2'-monophosphate (2'-AMP) is the 2'-monophosphate derivative of adenosine supplied as a research-grade biochemical reagent for studies of adenosine receptor signaling and nucleotide metabolism. The material is provided with batch purity information and is intended for laboratory research use only, not for human or diagnostic applications.
  • High purity suitable for analytical and biochemical assays.
  • Provided in multiple small pack sizes for low-quantity experiments.
  • Molecular formula C10H14N5O7P and molecular weight 347.22 g/mol.
  • Commonly used in adenosine receptor and nucleotide metabolism research.
  • Supplied with supporting product datasheet and batch-specific purity data.
Catalog No. 50-002-36995