Learn More
Medchemexpress LLC Adenosine, 5'-deoxy-5'-fluoro | 731-98-6 | Not available | 96.2% | 269.23 g/mol | C10H12FN5O3 | 50 MG

Supplier: Medchemexpress LLC HYW61673250MG
5′-Fluoro-5′-deoxyadenosine is a fluorinated adenosine nucleoside supplied as a research reagent for studies of nucleoside chemistry, enzymatic fluorination, and related in vitro and in vivo assays. It is intended for research use only.
- High purity (96.2%) suitable for biochemical assays.
- Molecular formula C10H12FN5O3; molecular weight 269.23 g/mol.
- CAS number 731-98-6 for unambiguous identification.
- Soluble in DMSO at 20 mg/mL (use freshly opened DMSO; adjust pH as directed).
- In vivo formulations supported: ≥2 mg/mL in 10% DMSO/40% PEG300/5% Tween-80/45% saline, or in 10% DMSO with 20% SBE-β-CD in saline.
- Storage: store at 4°C under nitrogen, protect from moisture; in solution store at -80°C up to 6 months or -20°C up to 1 month.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.