Learn More
Medchemexpress LLC Adenosine 5'-diphosphate (Standard) | 58-64-0 | 98.4% | 427.20 | 50 MG

Supplier: Medchemexpress LLC HYW010918R50MG
Adenosine 5'-diphosphate (Standard) is a laboratory chemical from MedChemExpress. It is a white solid that comes in a brown glass bottle and should be stored at 4°C, sealed, away from light and moisture. It is classified as harmful if swallowed, causes skin and serious eye irritation, and may cause respiratory irritation.
- Appearance: White solid
- Purity: 98.39% by HPLC
- Molecular formula: C10H15N5O10P2
- Molecular weight: 427.20
- CAS number: 58-64-0
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.