Learn More
Medchemexpress LLC Adenosine 5'-diphosphoribose | 20762-30-5 | 559.32 | C15H23N5O14P2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10097350MG
Encompass_Preferred
Adenosine 5'-diphosphoribose (ADP ribose) is a nucleotide metabolite supplied as a research reagent for biochemical and analytical studies. It is used to investigate NAD+ metabolism, TRPM2 channel activation, and ADP-ribosylation pathways. Intended for research use only.
  • Nucleotide metabolite for biochemical and analytical assays.
  • Molecular formula C15H23N5O14P2; molecular weight 559.32.
  • Used in studies of NAD+ metabolism, TRPM2 activation, and ADP-ribosylation.
  • Manufacturer datasheet lists pack sizes 1 mg, 5 mg, and 10 mg.
Catalog No. 50-002-12104