Learn More
Apexbio Technology LLC Adenosine 5-monophosphate 61-19-8 500mg
SDP
Supplier:  Apexbio Technology LLC B7875500
Encompass_Preferred
Adenosine 5 -monophosphate (AMP) is a nucleotide composed of the adenosine nucleoside esterified with a single phosphate group Serving as an intermediate in cellular energy metabolism AMP is involved in various biochemical pathways including the regulation of enzyme activity and signal transduction processes It plays a pivotal role in modulating the AMP-activated protein kinase (AMPK) pathway which governs cellular energy homeostasis AMP is widely utilized in research settings to study metabolic regulation nucleotide signaling and enzymatic reactions associated with phosphorylation and cellular energy status
Catalog No. 50-000-90885