Learn More
Medchemexpress LLC Adenosine 5'-monophosphate disodium | 4578-31-8 | 99.9% | 391.18 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01101210G
Encompass_Preferred
Adenosine 5'-monophosphate disodium is an orally active purine nucleotide that participates in ATP metabolism. It also serves as a ligand for the adenosine 2B receptor. This compound can activate AMPK in skeletal muscle, which helps to improve insulin resistance and impaired glucose metabolism, making it suitable for diabetes research.
  • Orally active purine nucleotide
  • Participates in ATP metabolism
  • Ligand for adenosine 2B receptor
  • Activates AMPK in skeletal muscle
  • Ameliorates insulin resistance and impaired glucose metabolism
  • Useful for research of diabetes
Catalog No. 50-003-66362