Learn More
Medchemexpress LLC HY-B0228 5g , Adenosine CAS:58-61-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB02285G
Encompass_Preferred
HY-B0228 5g Adenosine CAS: 58-61-7 Adenosine (Adenine riboside), a ubiquitous endogenous autacoid, acts through the enrollment of four G protein-coupled receptors: A1, A2A, A2B, and A3. Adenosine affects almost all aspects of cellular physiology, including neuronal activity, vascular function, platelet aggregation, and blood cell regulation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1123