Learn More
Genetex Inc Adenosine A1 Receptor Monoclonal Antibody [H341] HistoMAX™ (GTX641402) GeneTex - IHC-Optimized Antibody
SDP
Supplier:  Genetex Inc GTX641402
Encompass_Preferred
The Adenosine A1 Receptor antibody [H341] HistoMAX™ (GTX641402) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human normal tissues using GTX641402 Adenosine A1 Receptor antibody [H341] HistoMAX.™. A moderate to strong adenosine A1 receptor immunostaining can be found in the grey matter of the cerebrum and in the molecular layer of the cerebellum.
Catalog No. NC3669377