Learn More
Medchemexpress LLC Adenosine amine congener (ADAC) | 96760-69-9 | 99.2% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY128064100MG
Encompass_Preferred
Adenosine amine congener (ADAC) is a selective A1 adenosine receptor agonist known for its ability to ameliorate noise- and cisplatin-induced cochlear injury. It also exhibits neuroprotective effects, protecting sensory hair cells and reducing oxidative stress.
  • Selective A1 adenosine receptor agonist
  • Ameliorates noise- and cisplatin-induced cochlear injury
  • Provides neuroprotective effects
  • Reduces oxidative stress in the noise-exposed cochlea
  • Protects sensory hair cells and reduces cisplatin-induced apoptosis
  • Inhibits glutamate release and voltage-gated Ca2+ channels for otoprotection
Catalog No. 50-003-39218