Learn More
Medchemexpress LLC Adenosine antagonist-1 | 431040-19-6 | 99.8% | 359.41 g·mol⁻¹ | C18H13N7S | 10 MG

Supplier: Medchemexpress LLC HY10027410MG
Adenosine antagonist-1 is a small-molecule antagonist of the adenosine A3 receptor (A3R) reported to have anti-inflammatory activity. It is supplied as a white to off-white solid with high chemical purity and is intended for research use in studies of A3 receptor signaling and inflammatory pathways.
- High purity (99.8%).
- Molecular weight 359.41 g·mol⁻¹; formula C18H13N7S.
- White to off-white solid appearance.
- Storage: powder at -20°C; in solvent at -80°C (6 months) or -20°C (1 month).
- Suitable for research on A3 receptor-mediated inflammation and signaling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.