Learn More
Medchemexpress LLC Adenosine-d | 109923-50-4 | 99.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0228S1MG
Encompass_Preferred
Adenosine-d is a deuterium labeled Adenosine. Adenosine (Adenine riboside) is a ubiquitous endogenous autacoid that acts through four G protein-coupled receptors: A1, A2A, A2B, and A3. It affects almost all aspects of cellular physiology, including neuronal activity, vascular function, platelet aggregation, and blood cell regulation. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules for quantitation during drug development and can affect pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-05669