Learn More
Medchemexpress LLC Adenosine monophosphate-d12 dilithium | 61-19-8 | 95.7% | C10D12Li2N5O7P | 1 MG
Supplier: Medchemexpress LLC HYA0181S21MG
This product is a deuterium-labeled form of Adenosine monophosphate, which is recognized as an adenosine A1 receptor agonist. It demonstrates notable antiviral properties against both HSV-1 and HSV-2. Furthermore, it plays a critical role as a cellular metabolite, contributing to the regulation of energy balance and signal transmission within cells.
- Useful as a tracer in various research applications.
- Can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
- Incorporation of stable heavy isotopes (e.g., deuterium) can influence the pharmacokinetic and metabolic profiles of compounds, making it valuable for drug development.