Learn More
Medchemexpress LLC Adenosine, N-(1-oxobutyl)-, cyclic 3',5'-(hydrogen phosphate) 2'-butanoate, calcium salt (2:1) | 938448-87-4 | 98.58% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0764A10MM/1ML
Encompass_Preferred
Bucladesine calcium salt (Dibutyryl-cAMP calcium salt; DC2797 calcium salt) is a cell-permeable cyclic AMP (cAMP) analog that selectively activates cAMP dependent protein kinase (PKA) by increasing the intracellular level of cAMP. It also functions as a phosphodiesterase (PDE) inhibitor.
  • Cell-permeable cyclic AMP (cAMP) analog
  • Selectively activates cAMP dependent protein kinase (PKA)
  • Increases intracellular levels of cAMP
  • Functions as a phosphodiesterase (PDE) inhibitor
Catalog No. 50-004-08138