Learn More
Sigma Aldrich Fine Chemicals Biosciences Adenosne 35 dphosphate d100MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A5763100MG
Encompass
3AE-phosphoadenosine 5AE-phosphate (PAP) a 3AE-phosphorylated nucleotide is found in almost all organisms and is obtained as a by-product of sulfur and lipid metabolism.
Catalog No. 50-290-3094