Learn More
Medchemexpress LLC Adenosylcobalamin | 13870-90-1 | 99.2% | 1579.58 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11279010MM/1ML
Encompass_Preferred
Adenosylcobalamin (Coenzyme B12; Cobamamide; AdoCbl) is an active form of Vitamin B12 which is a cofactor for methylmalonyl CoA mutase.
  • Serves as a cofactor for a group of enzymes that catalyze unusual rearrangement or elimination reactions.
Catalog No. 50-004-37372