Learn More
Cell Biolabs Inc QuickTiter™ Adenovirus Quantitation Kit, 20 assays
Supplier:  Cell Biolabs Inc VPK106
Encompass_Preferred
Cell Biolabs' QuickTiter™ Adenovirus Quantitation Kit provides an ultra-fast 45-60 minute procedure that measures physical titer of purified virus or unpurified supernatant via the viral nucleic acid content. Quantitation is performed on a 96-well fluorescence based plate reader. The kit provides sufficient reagents for 20 assays.
Catalog No. NC9412849