Learn More
Cayman Chemical AdenylosuccInIc AcId 5mg
SDP
Supplier:  Cayman Chemical 296965 MG
Encompass_Preferred
A purine nucleotide and an intermediate in the purine nucleotide cycle; converted into AMP and fumarate by ADSL in the cytosol; inhibits calcium-induced activation of non-selective cation channels in isolated rat brown adipocytes in a patch-clamp assay at 10 µM; induces contractions in isolated guinea pig uterus strips at 100 µM; increases glucose-induced insulin exocytosis in INS-1 832/13 insulinoma cells at 10 µM; decreases muscle damage and pseudohypertrophy in a dystrophin-deficient mdx mouse model of DMD
Catalog No. 50-299-0278