Learn More
Medchemexpress LLC Adenylyl imidodiphosphate lithium hydrate | 94.0% | 506.20 g/mol (free acid) | C10H17N6O12P3·xLi·yH2O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY130777A5MG
Encompass_Preferred
AMP-PNP lithium hydrate (adenylyl imidodiphosphate lithium hydrate) is a non-hydrolyzable ATP analogue used to probe ATP-binding proteins and ATP-dependent processes. It competitively binds ATP sites without enzymatic hydrolysis, enabling controlled biochemical and biophysical studies of nucleotide-dependent systems.
  • Non-hydrolyzable ATP analog that competitively binds ATP sites.
  • Useful for enzyme assays, kinase regulation studies, and nucleotide metabolism research.
  • Supports investigations of ion channel function and protein complex assembly.
  • Supplied as a solid with high purity (94.0%).
  • Stable when stored sealed and protected from moisture under recommended temperatures.
  • Molecular weight 506.20 g/mol (free acid); formula C10H17N6O12P3·xLi·yH2O.
Catalog No. 50-002-62930