Learn More
Medchemexpress LLC HY-10293 5mg , Aderbasib CAS:791828-58-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY102935MG
Encompass_Preferred
HY-10293 5mg Aderbasib CAS: 791828-58-5 Aderbasib (INCB007839) is a potent, orally active and target specific low nanomolar hydroxamate-based inhibitor of ADAM10 and ADAM17. Aderbasib exhibits robust antineoplastic activity and can be used for cancer research, including diffuse large B-cell non-Hodgkin lymphoma, HER2+ breast cancer, gliomas, et al. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9903