Learn More
Cayman Chemical aDGalactopyrnosylphenyl Is 1mg
SDP
Supplier:  Cayman Chemical 195441 MG
Encompass_Preferred
Used to prepare various neoglycoproteins, which consist of a glycosylated serum albumin substituted with either fluorescein or methotrexate
Catalog No. 50-298-2689