Learn More
ABclonal Technology ADGRA3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A155731000UL
Encompass_Preferred
This gene encodes a member of the G protein-coupled receptor superfamily. This membrane protein may play a role in tumor angiogenesis through its interaction with the human homolog of the Drosophila disc large tumor suppressor gene. This gene is mapped to a candidate region of chromosome 4 which may be associated with bipolar disorder and schizophrenia.
Catalog No. 50-001-08260