Learn More
Medchemexpress LLC HY-13541 10mg , ADH-1 CAS:229971-81-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1354110MG
Encompass_Preferred
HY-13541 10mg ADH-1 CAS: 229971-81-7 ADH-1, an N-cadherin antagonist, inhibits N-cadherin mediated cell adhesion. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0074