Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ADH1B Human His-SU 5ug
SDP

Supplier:  Medchemexpress LLC HYP720715UG

Encompass_Preferred

Recombinant human ADH1B (alcohol dehydrogenase 1B) is expressed in Escherichia coli and supplied as an N-terminal 6xHis-SUMO-tagged protein. The product is lyophilized from PBS with 6% trehalose, shows >90% purity by reducing SDS-PAGE, and has an approximate molecular weight of 55.7 kDa. It is provided in microgram-scale quantities for biochemical and structural applications; aliquot after reconstitution and store frozen for long-term stability.

  • Recombinant human enzyme covering residues S2-F375 (UniProt P00325).
  • N-terminal 6xHis and SUMO tags facilitate purification and solubility.
  • Expressed in Escherichia coli for robust yield.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Lyophilized from PBS with 6% trehalose for enhanced stability.
  • Available in multiple microgram sizes suited to small-scale assays.
  • Store frozen; aliquot and keep at -80°C for long-term storage.

Catalog No. 50-001-96281


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.