Learn More
Med Vet International HoofGrip Block Adhesive 200ml, J1405, Each
Supplier:  Med Vet International JOR1405
Encompass_Preferred
Hoof Grip is a superior, two part urethane, fast-set adhesive used to bond blocks to bovine hooves. Our innovative cartridge can be closed airtight and stored for later use. Simply re-insert the same dual chamber plug removed to add the mixing tip. There is NO cutting required to open the new cartridge system. Is Hoof Grip Different? YES! n BONDS FASTER | BONDS STRONGER | NO WASTE NO MESS | EASE OF USE | MORE PRODUCT • 30 Second Fast Set • 200mL • No curing • Durable, Commercial Strength Cartridge • Dual Chamber Eliminates Cross Contamination • Made in the USA • Airtight Storage • Easy On & Off cap • No Harmful Odors • One Year Shelf Life
Catalog No. 50-004-95044