Learn More
Essendant ADHESIVEEPOXY.85OZ
Supplier:  Essendant GOR4200101
Encompass_Preferred
Gorilla takes epoxy to the Gorilla Tough level With its superior solvent and water resistance Gorilla Epoxy adhesive is incredibly strong and durable for household and automotive repairs alike The easy-to-use syringe keeps the epoxy resin and hardener separate so it is easy to dispense The two-part gap-filling formula easily bonds steel aluminum glass wood ceramic tile and most plastics Plus it dries in an ideal clear finish that makes projects look spotless Not recommended for use on polyethylene or polypropylene plastics Adhesive Type- Liquid Product Packaging- Syringe Washable/Removable- No Color on Application- Clear
Catalog No. 50-000-34792